Key Documents
R1138
Sigma-Aldrich
Rack for Nalgene® microcentrifuge tubes
Synonym(s):
microcentrifuge tube rack, microcentrifuge tube storage
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
material
polycarbonate
feature
Holds 81 x 0.5 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Nalgene® 5055-5005
size × H
5 1/4 in. × 5 1/4 in. × 2 in.
General description
Bottom of white polycarbonate, lid of clear polycarbonate, permanent foam insert.
For safe storage of samples in microcentrifuge tubes from −135 °C to +121 °C. Not for use in liquid-phase liquid nitrogen. May be repeatedly autoclaved. Holes in foam are placed diagonally and spaced to avoid overlapping of tube closures. Lid accepts writing with marker.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)