Skip to Content
MilliporeSigma
All Photos(1)

Documents

R1033

Sigma-Aldrich

RN-1747

≥98% (HPLC)

Synonym(s):

1-(4-Chloro-2-nitrophenyl)sulfonyl-4-benzylpiperazine;, 1-[(4-Chloro-2-nitrophenyl)sulfonyl]-4-(phenylmethyl)-piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18ClN3O4S
CAS Number:
Molecular Weight:
395.86
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to light tan

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cc(Cl)ccc1S(=O)(=O)N2CCN(CC2)Cc3ccccc3

InChI

1S/C17H18ClN3O4S/c18-15-6-7-17(16(12-15)21(22)23)26(24,25)20-10-8-19(9-11-20)13-14-4-2-1-3-5-14/h1-7,12H,8-11,13H2

InChI key

ZNLVYSJQUMALEO-UHFFFAOYSA-N

Biochem/physiol Actions

TRPV4, a close relative of the vanilloid receptor TRPV1, is activated by diverse modalities such as endogenous lipid ligands, hypotonicity, protein kinases and, possibly, mechanical inputs. TRPV4 is a nociceptor playing an important role in inflammatory and neuropathic mechanical hyperalgesia in rodent models apparently specifically activated under pathological conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osamu Nakanishi et al.
The Journal of pharmacology and experimental therapeutics, 373(2), 239-247 (2020-02-28)
Transient receptor potential (TRP) melastatin 8 (TRPM8) is a temperature-sensing ion channel mainly expressed in primary sensory neurons (Aδ-fibers and C-fibers in the dorsal root ganglion). In this report, we characterized KPR-5714 (N-[(R)-3,3-difluoro-4-hydroxy-1-(2H-1,2,3-triazol-2-yl)butan-2-yl]-3-fluoro-2-[5-(4-fluorophenyl)-1H-pyrazol-3-yl]benzamide), a novel and selective TRPM8 antagonist, to
Yousif A Shamsaldeen et al.
European journal of pharmacology, 888, 173441-173441 (2020-08-19)
Endothelial dysfunction is a common complication in diabetes in which endothelium-dependent vasorelaxation is impaired. The aim of this study was to examine the involvement of the TRPV4 ion channel in type 1 diabetic endothelial dysfunction and the possible association of
Mengqi Li et al.
Cell metabolism, 30(3), 508-524 (2019-06-18)
Fructose-1,6-bisphosphate (FBP) aldolase links sensing of declining glucose availability to AMPK activation via the lysosomal pathway. However, how aldolase transmits lack of occupancy by FBP to AMPK activation remains unclear. Here, we show that FBP-unoccupied aldolase interacts with and inhibits

Articles

We offer many products related to transient receptor potential channels for your research needs.

We offer many products related to transient receptor potential channels for your research needs.

We offer many products related to transient receptor potential channels for your research needs.

We offer many products related to transient receptor potential channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service