MilliporeSigma
  • Dysregulation of TRPV4, eNOS and caveolin-1 contribute to endothelial dysfunction in the streptozotocin rat model of diabetes.
Dysregulation of TRPV4, eNOS and caveolin-1 contribute to endothelial dysfunction in the streptozotocin rat model of diabetes.
European journal of pharmacology (2020-08-19)
Yousif A Shamsaldeen, Lisa A Lione, Christopher D Benham
ABSTRACT
Endothelial dysfunction is a common complication in diabetes in which endothelium-dependent vasorelaxation is impaired. The aim of this study was to examine the involvement of the TRPV4 ion channel in type 1 diabetic endothelial dysfunction and the possible association of endothelial dysfunction with reduced expression of TRPV4, endothelial nitric oxide synthase (eNOS) and caveolin-1. Male Wistar rats (350-450 g) were injected with 65 mg/kg i.p. streptozotocin (STZ) or vehicle. Endothelial function was investigated in aortic rings and mesenteric arteries using organ bath and myograph, respectively. TRPV4 function was studied with fura-2 calcium imaging in endothelial cells cultured from aortas from control and STZ treated rats. TRPV4, caveolin-1 and eNOS expression was investigated in these cells using immunohistochemistry. STZ-treated diabetic rats showed significant endothelial dysfunction characterised by impaired muscarinic-induced vasorelaxation (aortic rings: STZ-diabetics: Emax = 29.6 ± 9.3%; control: Emax = 77.2 ± 2.5% P˂0.001), as well as significant impairment in TRPV4-induced vasorelaxation (aortic rings, 4αPDD STZ-diabetics: Emax = 56.0 ± 5.5%; control: Emax = 81.1 ± 2.1% P˂0.001). Furthermore, STZ-diabetic primary aortic endothelial cells showed a significant reduction in TRPV4-induced intracellular calcium elevation (P˂0.05) compared with the control group. This was associated with significantly lower expression of TRPV4, caveolin-1 and eNOS and this was reversed by insulin treatment of the endothelial cultures from STZ -diabetic rats. Taken together, these data are consistent with the hypothesis that signalling through TRPV4, caveolin-1, and eNOS is downregulated in STZ-diabetic aortic endothelial cells and restored by insulin treatment.
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Endothelial cell growth supplement from bovine pituitary, ECGS, suitable for cell culture, BioReagent
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
RN-1747, ≥98% (HPLC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture