Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R0761

Sigma-Aldrich

Monoclonal Anti-Rat IgG2a antibody produced in mouse

clone R2A-2, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

R2A-2, monoclonal

contains

15 mM sodium azide

technique(s)

capture ELISA: suitable
dot blot: suitable
immunocytochemistry: suitable
indirect ELISA: 1:5,000
western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulins (IgGs) are glycoprotein antibodies that modulate several immune responses. Rat IgGs have four subclasses, namely, IgG1, IgG2a, IgG2b, and IgG2c.
Monoclonal Anti-Rat IgG2a (mouse IgG1 isotype) is derived from the R2A-2 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with purified rat IgG.

Specificity

Monoclonal Anti-Rat IgG2a recognizes an epitope located on the heavy chain of rat IgG2a. The antibody detects rat IgG2a derived from normal serum or myeloma proteins, but not the other rat immunoglobulins. Weak cross-reaction is observed with guinea pig immunoglobulins but not with IgG and serum preparations of bovine, cat, chicken, dog, goat, horse, human, mouse, pig, rabbit, or sheep.

Immunogen

Purified rat IgG

Application

Monoclonal Anti-Rat IgG2a antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • slot blot immunoassay
  • immunoblotting
  • dot blot
  • immunocytochemistry

Biochem/physiol Actions

Rat immunoglobulins (IgGs) against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rat IgG is a useful tool for the analysis of target proteins. Monoclonal Anti-Rat IgG2a antibody is specific for epitope on the heavy chain of rat IgG2a. The antibody localizes the denatured-reduced heavy chain molecule of rat IgG2a (γ2a). The product recognizes rat IgG2a derived from normal serum or myeloma proteins, but does not bind to other rat immunoglobulins. The antibody weakly associates with guinea pig immunoglobulins but does not react with IgG and serum preparations derived from bovine, cat, chicken, dog, goat, horse, human, mouse, pig, rabbit, and sheep.

Physical form

The product is provided as ascites fluid with 0.1% sodium azide as a preservative.

Storage and Stability

For continuous use, store at 0-5 °C. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use,

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three classes and four (sub)classes of rat immunoglobulins: IgM, IgA, IgE and IgG1, IgG2a, IgG2b, IgG2c.
H Bazin et al.
European journal of immunology, 4(1), 44-48 (1974-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service