Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0412

Sigma-Aldrich

PF-06427878

≥98% (HPLC)

Synonym(s):

(R)-3-((2-(3-(2-Ethoxyphenoxy)piperidin-1-yl)pyrimidine-5-carboxamido)methyl)benzoic acid, 3-[[[[2-[(3R)-3-(2-Ethoxyphenoxy)-1-piperidinyl]-5-pyrimidinyl]carbonyl]amino]methylbenzoic acid, PF 06427878, PF06427878

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28N4O5
CAS Number:
Molecular Weight:
476.52
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(C1=CC=CC(CNC(C2=CN=C(N=C2)N3C[C@@H](CCC3)OC4=CC=CC=C4OCC)=O)=C1)O

InChI

1S/C26H28N4O5/c1-2-34-22-10-3-4-11-23(22)35-21-9-6-12-30(17-21)26-28-15-20(16-29-26)24(31)27-14-18-7-5-8-19(13-18)25(32)33/h3-5,7-8,10-11,13,15-16,21H,2,6,9,12,14,17H2,1H3,(H,27,31)(H,32,33)/t21-/m1/s1

InChI key

IDVOZBCJUVAIMF-OAQYLSRUSA-N

Biochem/physiol Actions

PF-06427878 is an orally active, potent and selective diacylglycerol acyltransferase 2 inhibitor (human/rat DGAT2 IC50 = 99/202 nM; >470-fold selectivity over DGAT1, MGAT1/2/3) that inhibits DGAT2-dependent triglyceride (TG) synthesis in primary human hepatocytes (IC50 = 11.6 nM in the presence of DGAT1 inhibitor PF-04620110). PF-06427878 reduces hepatic and circulating plasma TG levels as well as lipogenic gene expression in rats maintained on a Western-type diet (0.3-30 mg/kg bid. po.). Likewise, PF-06427878 significantly improves steatosis and hepatocellular ballooning with a decrease in lobular inflammation in a murine nonalcoholic steatohepatitis (NASH) model (2 or 20 mg/kg bid. po.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neeta B Amin et al.
Science translational medicine, 11(520) (2019-11-30)
Nonalcoholic steatohepatitis (NASH) is characterized by the accumulation of hepatocyte triglycerides, the synthesis of which is catalyzed by diacylglycerol acyltransferases (DGATs). Here, we investigate DGAT2 as a potential therapeutic target using an orally administered, selective DGAT2 inhibitor, PF-06427878. Treatment with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service