Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0328

Sigma-Aldrich

PF-4800567 hydrochloride

≥98% (HPLC)

Synonym(s):

3-[(3-Chlorophenoxy)methyl]-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine hydrochloride, PF 4800567

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18ClN5O2 · HCl
CAS Number:
Molecular Weight:
396.27
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

shipped in

dry ice

storage temp.

−20°C

SMILES string

Clc1cc(ccc1)OCc2n[n](c4ncnc(c42)N)C3CCOCC3.Cl

InChI

1S/C17H18ClN5O2.ClH/c18-11-2-1-3-13(8-11)25-9-14-15-16(19)20-10-21-17(15)23(22-14)12-4-6-24-7-5-12;/h1-3,8,10,12H,4-7,9H2,(H2,19,20,21);1H

InChI key

QZXZQMUZEHTFHD-UHFFFAOYSA-N

Biochem/physiol Actions

PF-4800567 is a casein kinase 1e (CK1e) selective inhibitor (IC50 = 32 nM) that is 20 fold selective for the CK1e isoform over CK1d. PF-4800567 blockes CK1e-mediated PER3 nuclear translocation, but does not effect the circadian clock in animal studies.
PF-4800567 is also known as (3-[(3-Chlorophenoxy)methyl]-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine hydrochloride). It can block the epidermal growth factor receptor (EGFR) at micromolar concentrations. PF-4800567 helps to improve the locomotor stimulant reaction to methamphetamine (MA) and fentanyl.

Other Notes

PF-4800567 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the PF-4800567 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Csnk1e is a genetic regulator of sensitivity to psychostimulants and opioids
Bryant, et al.
Neuropsychopharmacology, 37(4), 1026-1026 (2012)
Combined Pharmacological and Genetic Manipulations Unlock Unprecedented Temporal Elasticity and Reveal Phase-Specific Modulation of the Molecular Circadian Clock of the Mouse Suprachiasmatic Nucleus.
Patton AP, et al.
The Journal of Neuroscience, 36(36), 9326-9341 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service