Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0281

Sigma-Aldrich

PF-05212384

≥98% (HPLC)

Synonym(s):

1-(4-((4-(Dimethylamino)piperidin-1-yl)carbonyl)phenyl)-3-(4-(4,6-dimorpholin-4-yl-1,3,5-triazin-2-yl)phenyl)urea, Gedatolisib, PKI-587

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H41N9O4
CAS Number:
Molecular Weight:
615.73
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 0.5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

N6(CCOCC6)c1nc(nc(n1)c3ccc(cc3)NC(=O)Nc4ccc(cc4)C(=O)N5CCC(CC5)N(C)C)N2CCOCC2

InChI

1S/C32H41N9O4/c1-38(2)27-11-13-39(14-12-27)29(42)24-5-9-26(10-6-24)34-32(43)33-25-7-3-23(4-8-25)28-35-30(40-15-19-44-20-16-40)37-31(36-28)41-17-21-45-22-18-41/h3-10,27H,11-22H2,1-2H3,(H2,33,34,43)

InChI key

DWZAEMINVBZMHQ-UHFFFAOYSA-N

Biochem/physiol Actions

PF-05212384, also known as PKI-587 and Gedatolisib, is a dual phosphatidylinositol-3-kinase (PI3K) / mammalian target of rapamycin (mTOR) inhibitor. PKI-587 potently inhibited class I PI3Ks (IC50 vs PI3K-α = 0.4 nM), PI3K-α mutants, and mTOR. PF-05212384 inhibited growth of 50 diverse human tumor cell lines at IC50s <100 nM and has been in clinical trials in patients with advanced solid tumors. PF-05212384 has also been found to sensitize cells to radiation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service