Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0303

Sigma-Aldrich

PF-04449913 maleate salt

≥98% (HPLC)

Synonym(s):

1-((2R,4R)-2-(1H-benzo[d]imidazol-2-yl)-1-methylpiperidin-4-yl)-3-(4-cyanophenyl)urea maleate salt, Glasdegib maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N6O · C4H4O4
CAS Number:
Molecular Weight:
490.51
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

OC(/C=C\C(O)=O)=O.CN1CC[C@@H](NC(NC2=CC=C(C#N)C=C2)=O)C[C@@H]1C3=NC4=CC=CC=C4N3

InChI

1S/C21H22N6O.C4H4O4/c1-27-11-10-16(24-21(28)23-15-8-6-14(13-22)7-9-15)12-19(27)20-25-17-4-2-3-5-18(17)26-20;5-3(6)1-2-4(7)8/h2-9,16,19H,10-12H2,1H3,(H,25,26)(H2,23,24,28);1-2H,(H,5,6)(H,7,8)/b;2-1-/t16-,19-;/m1./s1

InChI key

VJCVKWFBWAVYOC-UIXXXISESA-N

Gene Information

human ... SMO(6608)

Biochem/physiol Actions

PF-04449913 (Glasdegib) is an orally bioavailable Hedgehog pathway inhibitor. PF-04449913 acts by binidng Smoothened (Smo) and blocking signal transduction. PF-04449913 has an IC50 of 5 nM in the Gli-Luciferase assay. In a Phase I clinical trial in patients with advanced solid tumors, PF-04449913 caused disease stabilization in 34% of patients.
PF-04449913 maleate salt is also known as 1-((2R,4R)-2-(1H-benzo[d]imidazol-2-yl)-1-methylpiperidin-4-yl)-3-(4-cyanophenyl)urea. It increases the differentiation of blood cells from hematopoietic precursors of the lymph gland medullary zone. This human Smo inhibitor protein is now under clinical trials to treat myeloid malignancies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The human Smoothened inhibitor PF-04449913 induces exit from quiescence and loss of multipotent drosophila hematopoietic progenitor cells.
Giordani G, et al.
Oncotarget, 7(34), 55313-55313 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service