Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0273

Sigma-Aldrich

PF-04531083

≥98% (HPLC)

Synonym(s):

N-[6-Amino-5-(2-chloro-5-methoxyphenyl)-2-pyridinyl]-1-methyl-1H-pyrazole-5-carboxamide, N-[6-Amino-5-(2-chloro-5-methoxyphenyl)pyridin-2-yl]-1-methyl-1H-pyrazole-5-carboxamide, PF 4531083

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16ClN5O2
CAS Number:
Molecular Weight:
357.79
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

Clc1c(cc(cc1)OC)c2c(nc(cc2)NC(=O)c3[n](ncc3)C)N

InChI

1S/C17H16ClN5O2/c1-23-14(7-8-20-23)17(24)22-15-6-4-11(16(19)21-15)12-9-10(25-2)3-5-13(12)18/h3-9H,1-2H3,(H3,19,21,22,24)

InChI key

ZRJGMDIPCQOGNI-UHFFFAOYSA-N

Biochem/physiol Actions

PF-04531083 is an orally available, potent and selective sodium channel Nav1.8 blocker. PF-04531083 was designated as a potential treatment of pain.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service