Skip to Content
MilliporeSigma

D5220

Doxorubicin Ready-Made

2mg/ml, Suitable for cell culture

Synonym(s):

Adriamycin, DOX, Hydroxydaunorubicin hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.76
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

mol wt

579.98 Da

packaging

pkg of 1 mL

concentration

2 mg/mL

shipped in

dry ice

storage temp.

-10 to -25°C

Biochem/physiol Actions

Doxorubicin is an antitumor antibiotic that acts by inhibiting DNA topoisomerase II. It intercalates with DNA and induces apoptosis by blocking replication.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library