Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PRS2325

Sigma-Aldrich

Anti-CASPASE-12 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Casp-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 46 kDa

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Casp12(12364)

General description

Caspase 12 (CASP12), a member of the caspase family, is expressed in the Golgi complex.

Immunogen

a peptide corresponding to amino acids 2-17 of murine caspase-12.

Application

Anti-CASPASE-12 (ab1) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Murine caspase 12 (CASP12) is activated during endoplasmic reticulum stress and activates other caspases which act on substrates, ultimately leading to apoptosis. The protein has been shown to be activated during progression of retinal degeneration in animal models. In mice models, CASP12 has been shown to cause neuronal death resulting from ischemia/reperfusion.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2325.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ameliorating effect of Klotho on endoplasmic reticulum stress and renal fibrosis induced by unilateral ureteral obstruction
Qi-Feng
Iranian Journal of Kidney Diseases (2015)
Targeting Caspase-12 to Preserve Vision in Mice With Inherited Retinal Degeneration
Yogesh B
Investigative Ophthalmology & Visual Science (2015)
Activation of caspase-12 by endoplasmic reticulum stress induced by transient middle cerebral artery occlusion in mice.
Shibata M
Neuroscience (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service