Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0184

Sigma-Aldrich

Rabbit anti-PARP1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADP-ribosyltransferase NAD(+), ADP-ribosyltransferase diphtheria toxin-like 1, ADPRT, ADPRT 1, ADPRT1, ARTD1, NAD(+) ADP-ribosyltransferase 1, PARP, PARP-1, PPOL, member 1, pADPRT-1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, poly[ADP-ribose] synthase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

western blot: 1:2,000-1:10,000

accession no.

NP_001609.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PARP1(142)

Immunogen

The epitope recognized by PLA0184 maps to a region between residue 1 and 50 of human poly (ADP-ribose) polymerase family, member 1 using the numbering given in entry NP_001609.1 (GeneID 142).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

PARP1 (Poly-ADP-ribose polymerase 1) is a member of PARP family of enzymes that transfer the ADP-D-ribosyl group of NAD(+) to an acceptor carboxyl group on proteins. PARP1 catalyzes the poly-ADP-ribosylation of histone and non-histone proteins in response to DNA damage. The over-activation of PARP-1 in response to DNA damage has been shown to promote cell and tissue injury. This observation has initiated research into the therapeutic potential of PARP1 inhibitors in the treatment of cancer, cardiovascular disease, and brain injury.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim-Ling Chin et al.
Scientific reports, 12(1), 1054-1054 (2022-01-22)
Neonatal microcephaly and adult Guillain-Barré syndrome are severe complications of Zika virus (ZIKV) infection. The robustly induced inflammatory cytokine expressions in ZIKV-infected patients may constitute a hallmark for severe disease. In the present study, the potential role of high mobility
Devon Lundine et al.
Molecular cancer research : MCR, 20(12), 1799-1810 (2022-09-09)
The TP53 gene is mutated in 80% of triple-negative breast cancers. Cells that harbor the hot-spot p53 gene mutation R273H produce an oncogenic mutant p53 (mtp53) that enhances cell proliferative and metastatic properties. The enhanced activities of mtp53 are collectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service