Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9041

Sigma-Aldrich

Phosphodiesterase II from bovine spleen

lyophilized powder, ≥5.0 units/mg protein

Synonym(s):

3′-Exonuclease, Spleen exonuclease, Spleen phosphodiesterase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥5.0 units/mg protein

mol wt

65 kDa

UniProt accession no.

foreign activity

5′-Nucleotidase ≤1% of base activity

storage temp.

−20°C

Gene Information

General description

Research area: Cell signaling. Phosphodiesterase (PDE) II belongs to the PDE superfamily and includes subtypes from one to 11. They exist as a dimer and comprise of N-terminal regulatory domain, C-terminal prenylated domain, and a Zn2+containing catalytic domain. The bovine spleen PDE2 corresponds to a molecular weight of 65 kDa. It requires divalent cations for its enzymatic activity and has an optimum pH of 7.5.

Application

Phosphodiesterase (PDE) is any enzyme that is used to break phosphodiester bonds. It is a membrane-bound glycoprotein that is used to catalyze the hydrolysis of various nucleotide polyphosphates. Phosphodiesterase II has been used in the enzymatic digestions of purified proteins such as the P8-dGMP complex. Bovine spleen phosphodiesterase has been used to digest N-cadherin. Phosphodiesterase II from bovine is suitable for sequential and end-group analysis of poly and oligonucleotides. It may be used in kinetic studieson substrate degradation of macro-molecules.
Phosphodiesterase II from bovine spleen has been used in the:
  • excision of pyridyloxobutyl (POB) base adducts from DNA
  • digestion of DNA prior to cycloadenosine enrichment
  • hydrolysis of DNA from blue mussels gill tissue into deoxyribonucleoside 3′-monophosphates

The product has been used in the characterization of polynucleotide chain length, base composition, and identity of terminal nucleotide. The enzyme has also been used in excision of pyridyloxobutyl (POB) base adducts from DNA. Furthermore, it has been used along with micrococcal endonuclease to hydrolyze purified DNA to 3 -nucleoside monophosphates.

Biochem/physiol Actions

Phosphodiesterase (PDE) breaks phosphodiester bonds. 3-Isobutyl-methylxanthine (IBMX) is a potential PDE2 inhibitor.
The enzyme acts on poly(A), poly(U), and poly(I). Native DNA and poly(C) are quite resistant to the action of this enzyme.
Hydrolyzes RNA, RNA-Core, 3′-alkyl- and 3′-aryl-nucleoside phosphates, and polydeoxyribonucleotides with 3′-phosphate end groups to 3′-mononucleotides.
Polynucleotides having 5′-phosphomonoester end groups are not attacked.

Unit Definition

One unit will produce acid soluble nucleotides equivalent to a ΔA260 of 16 in 30 min at pH 6.5 at 37 °C, in a 2.0 mL reaction mixture. Substrate: RNA-Core. Actual A260 is measured on the supernatant after precipitation of the unhydrolyzed RNA with uranyl acetate-perchloric acid reagent.

Analysis Note

Protein determined by biuret

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Hong Feng et al.
Biochemistry, 44(34), 11486-11495 (2005-08-24)
Endonuclease V is an enzyme that initiates a conserved DNA repair pathway by making an endonucleolytic incision at the 3' side one nucleotide from a deaminated base lesion. Site-directed mutagenesis analysis was conducted at seven conserved motifs of the thermostable
D H Bamford et al.
Journal of virology, 50(2), 309-315 (1984-05-01)
DNA of bacteriophage PRD1 has protein P8 at its termini. Extracts of infected cells are able to derivatize P8 in vitro with labeled dGTP. Two early proteins, P1 and P8, products of genes I and VIII, respectively, are the only
Dynamic regulation of cAMP signaling by cGMP in the cardiovascular system: roles of phosphodiesterase 2 and phosphodiesterase 3 enzymes.
Donald H Maurice
Proceedings of the Western Pharmacology Society, 46, 32-36 (2004-01-01)
Ekaterina M Fock et al.
Pflugers Archiv : European journal of physiology, 448(2), 197-203 (2004-01-15)
The present study addressed the question of whether nitric oxide (NO) participates in regulation of osmotic water permeability in the urinary bladder of the frog Rana temporaria L. Experiments were carried out on isolated, paired hemi-bladders filled with amphibian Ringer
Characterization of novel phosphodiesterases in the bovine ovarian follicle
Sasseville M, et al.
Biology of Reproduction, 81(2), 415-425 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service