Key Documents
P7049
Finnpipette® pipette tips
Finntip 250 universal, non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
yellow
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 ea (Racks)
manufacturer/tradename
Finn 9400260
L
5.2 cm
maximum volume
250 μL
volume range
0.5-250 μL
General description
Each tray contains 8 × 12 (96) tips. When used with multi-channel pipettes, spacing between tips in tray permits mounting up to 12 tips at once.
Other Notes
Browse all pipette tips
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries