Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7036

Sigma-Aldrich

Parathyroid hormone human

≥92% (HPLC), powder

Synonym(s):

PTH, Parathormone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C408H674N126O126S2
CAS Number:
Molecular Weight:
9424.62
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder
Assay:
≥92% (HPLC)

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥92% (HPLC)

form

powder

composition

Peptide content, ≥70%

solubility

water: 1 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... PTH(5741)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-Val-Ala-Leu-Gly-Ala-Pro-Leu-Ala-Pro-Arg-Asp-Ala-Gly-Ser-Gln-Arg-Pro-Arg-Lys-Lys-Glu-Asp-Asn-Val-Leu-Val-Glu-Ser-His-Glu-Lys-Ser-Leu-Gly-Glu-Ala-Asp-Lys-Ala-Asp-Val-Asn-Val-Leu-Thr-Lys-Ala-Lys-Ser-Gln

General description

Parathyroid hormone (PTH) is a circulating hormone secreted from the parathyroid gland. This hormone is synthesized as a pre-pro hormone. PTH is composed of binding regions that have a high affinity to a receptor and functional domains that interact with the receptors to initiate an intracellular signaling cascade.

Application

Parathyroid hormone human has been used to incubate mature human adipocytes.

Biochem/physiol Actions

Parathyroid hormone (PTH) exerts its effects on bone and kidney cells. This hormone plays a role in calcium homeostasis. PTH is secreted from the chief cells of the parathyroid gland in response to low extracellular calcium levels or higher levels of extracellular phosphate. PTH deficiency or excess PTH are associated with hypoparathyroidism and hyperparathyroidism, respectively.

Components

Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-Val-Ala-Leu-Gly-Ala-Pro-Leu-Ala-Pro-Arg-Asp-Ala-Gly-Ser-Gln-Arg-Pro-Arg-Lys-Lys-Glu-Asp-Asn-Val-Leu-Val-Glu-Ser-His-Glu-Lys-Ser-Leu-Gly-Glu-Ala-Asp-Lys-Ala-Asp-Val-Asn-Val-Leu-Thr-Lys-Ala-Lys-Ser-Gln

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Breining et al.
European journal of endocrinology, 184(5), 687-697 (2021-03-09)
Activation of brown adipose tissue is a promising strategy to treat and prevent obesity and obesity-related disorders. Activation of uncoupling protein 1 (UCP1) leads to uncoupled respiration and dissipation of stored energy as heat. Induction of UCP1-rich adipocytes in white

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service