Skip to Content
MilliporeSigma

L6420

Luteinizing Hormone from human pituitary

≥8,500 IU/mg

Synonym(s):

ICSH, LH, PLH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
254-420-6
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

specific activity

≥8,500 IU/mg

shipped in

dry ice

storage temp.

−20°C

Biochem/physiol Actions

Two-chain glycoprotein hormone; the α-chain is identical to that of chorionic gonadotropin, TSH, and FSH, while biological specificity is attributed to the distinct β-chain. Induces ovulation, spermatogenesis, and synthesis of steroid sex hormones.
Two-chain glycoprotein hormone; the α-chain is identical to that of chorionic gonadotropin, TSH, and FSH, while biological specificity is attributed to the distinct β-chain. Induces ovulation, spermatogenesis, and synthesis of steroid sex hormones. Activity is mediated through a trans-membrane LH/chorionic gonadotropin receptor that is a member of the G-protein coupled receptor (GPCR) family.

Physical form

Lyophilized from 50 mM ammonium bicarbonate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library