Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

P5498

Sigma-Aldrich

Anti-Pro-NGF antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Pro-Nerve Growth Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
western blot: 1:200-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NGFR(4804)
mouse ... Ngfr(18053)
rat ... Ngfr(24596)

Related Categories

Specificity

Does not cross react with pro-BDNF, pro-NT-3, or mature NGF.

Immunogen

peptide corresponding to amino acid residues 84-104 of rat NGF (precursor). The sequence is identical in mouse and 16/21 residues identical in human.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, 1% BSA and <0.1% sodium azide

Preparation Note

Reconstitute the lyophilized vial with 0.2 ml deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Zhang et al.
Molecular neurobiology, 58(9), 4237-4250 (2021-05-10)
Maternal subclinical hypothyroidism (SCH) during pregnancy can adversely affect the neurodevelopment of the offspring. The balance of nerve growth factor (NGF)-related tropomyosin receptor kinase A/p75 neurotrophin receptor (TrkA/p75NTR) signaling in the hippocampus is important in brain development, and whether it
Sabah Shiri et al.
RSC advances, 9(67), 38912-38927 (2019-11-27)
Drug delivery systems have been of interest to researchers. The effects of synthesized nano-polymers as silibinin and silymarin extract drug delivery systems on olfactory ensheathing cells under normal and high-glucose conditions were studied. The structure of the nanopolymer was characterized
Elahe Karimi et al.
Drug design, development and therapy, 13, 2513-2532 (2019-08-24)
Nowadays, the drug delivery system is important in the treatment of diseases. A polymeric nanoparticle modified by oleic acid (NPMO) as a Thymol (Thy) drug release system was synthesized from Thymbra spicata and its neurotrophic and angiogenic effects on rat's
Li Duan et al.
PloS one, 8(9), e73768-e73768 (2013-09-17)
While aberrant activation of microglial cells was evidently involved in neuroinflammation and neurotoxicity in the neurodegenerative diseases such as Alzheimer's and Parkinson's disease, objective of study was to address if activated microglias deliver their effect by releasing pro-neurotrophins. By in
Yi Xia et al.
International journal of molecular sciences, 14(7), 14085-14104 (2013-07-25)
Growing evidence has shown that proNGF-p75NTR-sortilin signaling might be a crucial factor in neurodegeneration, but it remains unclear if it may function in nigral neurons under aging and disease. The purpose of this study is to examine and quantify proNGF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service