Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3921

Sigma-Aldrich

Parathyroid Hormone Fragment 1-34 rat

≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C180H291N55O48S2
CAS Number:
Molecular Weight:
4057.71
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Pth(24694)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-leu-Ala-Ser-Val-Glu-Arg-Met-Gln-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe

Application

Parathyroid Hormone Fragment 1-34 rat has been used to study the in vivo effect of parathyroid hormone (PTH) on the intestinal HCO3-secretion in rats.[1] It has also been injected into rats to increase PTH levels and study its effect on the high-phosphate diet-induced elevation of blood pressure.[2]

Biochem/physiol Actions

Parathyroid Hormone Fragment 1-34 is a parathyroid hormone (PTH) analog. It is implicated in the stimulation of intracellular cAMP accumulation and it induces osteocalcin (OC) promoter activity.[3]

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X P Yu et al.
Endocrinology, 138(8), 3085-3092 (1997-08-01)
Osteocalcin (OC) is a bone-specific extracellular matrix protein expressed by mature osteoblasts during late stages of differentiation. Previous studies have shown that forskolin, an activator of adenylate cyclase, stimulated OC production. Because PTH has been shown to activate several intracellular
Narattaphol Charoenphandhu et al.
Biochemical and biophysical research communications, 409(4), 775-779 (2011-05-31)
Parathyroid hormone (PTH) was recently demonstrated to enhance the HCO(3)(-) secretion through the apical anion channel, cystic fibrosis transmembrane conductance regulator (CFTR), but how the HCO(3)(-) entered the epithelial cells was not well understood, in part, due to the lack
Milica Bozic et al.
Journal of hypertension, 32(9), 1822-1832 (2014-07-01)
There is growing evidence suggesting that phosphate intake is associated with blood pressure levels. However, data from epidemiological studies show inconsistent results. The present study was designed to evaluate the effect of high circulating phosphorus on arterial blood pressure of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service