Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P350-05

Sigma-Aldrich

Porcine Coronary Artery Smooth Muscle Cells: PCASMC (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

Porcine coronary artery

Quality Level

form

solid

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 38

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

diabetes; stroke; cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


PCASMC were used to investigate the role of c-Jun in smooth muscle proliferation, and to show that c-Jun inhibition abrogates smooth muscle cell growth and intimal thickening after arterial injury (Khachigian, 2002). PCASMC were also used to develop poly (l-lactide-co-caprolactone) nanofibrous scaffold material with potential for tissue engineering (Dong, 2008).

Characterization: positive for smooth muscle cell specific alpha-actin expression.

Cell Line Origin

Artery

Application

vascular research, supply of blood to heart muscle

Components

Porcine Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Porcine Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the PCASMC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Young Choi et al.
Biomaterials, 269, 120222-120222 (2020-08-02)
Stem cell fate is largely determined by cellular signaling networks and is heavily dependent on the supplementation of exogenous recombinant proteins into culture media; however, uneven distribution and inconsistent stability of recombinant proteins are closely associated with the spontaneous differentiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service