Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P300-05

Sigma-Aldrich

Porcine Coronary Artery Endothelial: PCAEC (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

Porcine coronary artery

Quality Level

form

solid

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 38

morphology

Endothelial

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Arterial endothelial cells play critical roles in cardiac homeostasis, and insults to the endothelial cells have been linked to a variety of vascular diseases. Porcine artery endothelial cells provide a useful tool to study artery endothelial function and metabolism.PCAEC from Cell Applications, Inc. have been used to investigate a new mechanism of obesity-associated vascular disease by demonstrating that resistin, an adipocyte-derived cytokine implemented in insulin resistance and diabetes, can cause endothelial dysfunction in coronary arteries through oxidative stress and down-regulation of NO synthase (Kougias, 2005). Conversely, secretoneurin was shown to activate expression of NO synthase and calmodulin, leading to endothelial relaxation (Chan, 2011). PCAEC were also used in adhesion and transmigration experiments in a study investigating anti-inflammatory, cardioprotective effects of a novel adenosine-lidocaine drug formulation (Shi, 2012).

Cell Line Origin

Artery

Application

cardiovascular function, angiogenesis, vascular biology, regulation of coronary blood flow, cardiac functions, effects of vaso-dilators, coagulation, fibrinolysis, vessel wall tone, cytokine production

Components

Porcine Endothelial Cell Growth Medium that contains 10% FBS and 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Porcine Endothelial Cell Growth Medium that contains 10% FBS and 10% DMSO
  • Can be cultured at least 12 doublings

Subculture Routine

Please refer to the PCAEC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service