Key Documents
OH30378543
Ohaus® Dissolved Oxygen Meter
Model ST400D
About This Item
UNSPSC Code:
41115611
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30378543
dissolved oxygen measuring range
0.00-20.0 mg/L (0 - 50 °C)
battery power
4 AAA, 250 hr life
General description
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results.
Large backlit LCD screen provides easy to read results - even in low light environments. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
Application
Field and lab Dissolved Oxygen measurement
Features and Benefits
99 measurement memory, last calibration data recall, auto barometric pressure measurement, auto temperature compensation with manual salinity compensation
Legal Information
Ohaus is a registered trademark of Ohaus Corp.