Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OH30129896

Ohaus® pH Meter

Model ST5000-F, AC/DC input 120 V, US 3-pin plug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

material

epoxy shaft

feature

CSA approved

packaging

pack of 1 ea

manufacturer/tradename

Ohaus® 30129896

ORP measuring range

-2000.00-+2000.00 mV

pH measuring range

-2-20 pH unit

temperature measuring range

-30-130 °C

Looking for similar products? Visit Product Comparison Guide

General description

Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
The color touchscreen operates similar to smartphones and tablet computers and provides simple operation of the advanced features, such as one-touch toggling of measurement modes.

Application

pH, oxidation-reduction potential (ORP) measurement

Features and Benefits

3 endpoint modes, continuous measurement mode, up to 9 point calibration, 1,000 measurement memory
Automatic and manual temperature compensation, auto buffer recognition, calibration icon

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service