Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z527807

Sigma-Aldrich

Oakton® Acorn series pH 6 meter

reads pH, mV, and temperature

Synonym(s):

Oakton pH meter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.79

feature

CE compliant

manufacturer/tradename

Oakton

pH measuring range

0.00-14.00 pH unit, accuracy: 0.01 pH unit, resolution: 0.01 pH unit

temperature measuring range

0.0-100.0 °C, accuracy: 0.5 °C, resolution: 0.1 °C

weight

1 lb

General description

  • Reads pH, mV, and temperature
  • Three-point pH calibration w/auto buffer recognition
  • Automatic Temperature Compensation (ATC)
  • Toggle between pH and temperature in °C
  • Push-button operation; hold function; auto-off
  • 3-year warranty on meter

Meter includes:
  • pH electrode w/BNC connector, temperature probe, operating instructions, and batteries.
  • pH calibration up to 3 points: 4.01; 7.00; 10.01 increments.

Temperature calibration:
  • Offset 0.1°C increments
  • mV calibration: ±20 mV

Legal Information

Acorn is a trademark of Cole-Parmer Instrument Co.
Oakton is a registered trademark of Cole-Parmer Instrument Co.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service