Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N6905

Sigma-Aldrich

NZCYM Broth

EZMix powder microbial growth medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

sterility

non-sterile

form

powder

application(s)

food and beverages

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

NZCYM is a microbial growth medium used for the cultivation of E. coli and lambda bacteriophage. This nutrient-rich microbial broth contains peptides, amino acids, and water-soluble vitamins in a low-salt formulation.

Features and Benefits

NZCYM EZMix powder provides:
  • Granulated, dust-free format for safer handling and faster mixing
  • Easy scale-up using larger package size
  • Standard formulation

Components

10 g/L Tryptone
5 g/L Yeast Extract
5 g/L NaCl
1 g/L Casamino Acids
1 g/L MgSO4
0.6 g/L inert binder

Reconstitution

Stir to suspend 22.6 g powder in 1L water. Autoclave for 15 minutes at 121°C to sterilize. Allow to cool before making additions, such as antibiotics (if desired).

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method for bacteriophage isolation against target Campylobacter strains
C. Carvalho
Letters in Applied Microbiology (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service