Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N0186

Sigma-Aldrich

Nourseothricin sulfate

powder, suitable for cell culture, BioReagent

Synonym(s):

ClonNAT, NTC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.76

Product Name

Nourseothricin sulfate, suitable for cell culture

biological source

Streptomyces sp.

Quality Level

form

powder

storage temp.

2-8°C

General description

Chemical structure: peptidyl nucleoside, aminoglycoside antibiotic. Nourseothricin (NTC) is a mixture of the related compounds streptothricin C, D, E, and F.

Application

Nourseothricin is used as a dominant selection antibiotic for genetically modified bacteria, yeasts, fungi, protozoa and plants.

Biochem/physiol Actions

Nourseothricin inhibits biosynthesis and induces miscoding. Resistance to nourseothricin is mediated by the sat1 encoded N-acetyltransferase. Nourseothricin is inactivated by acetylation of the β-amino group of the β-lysin.

Antifungal effective against Candida albicans. Candida species transformed with the gene encoding nourseothricin acetyltransferase (CaNAT1) were resistant to nourseothricin.

Packaging

10mg, 50mg

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service