Skip to Content
MilliporeSigma
All Photos(1)

Documents

N5540

Sigma-Aldrich

Nitrate Reductase (NAD[P]H) From Pichia Pastoris, recombinant

freeze-dried protein glass

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in Pichia pastoris

Quality Level

form

freeze-dried protein glass

specific activity

20-40 units/mg protein

storage temp.

−20°C

Biochem/physiol Actions

Catalyzes the NADH or NADPH dependent reduction of nitrate to nitrite.

Unit Definition

Unit Definition: One unit will reduce 1.0 μmole of nitrate to nitrite per minute in an NADH system at pH 7.5 at 30°C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service