Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10128023001

Roche

NADH

Grade II, disodium salt

Synonym(s):

β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, β-DPNH, β-NADH, DPNH, Diphosphopyridine nucleotide, reduced form, NADH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O14P2 · xH2O
CAS Number:
Molecular Weight:
709.40 (anhydrous basis)
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Quality Level

Assay

98%

form

amorphous powder

mol wt

(NADH-Na2: Mr = 709.4
NADH: Mr = 665.4)

packaging

pkg of 1 g

manufacturer/tradename

Roche

storage condition

protect from light

λmax

260 and 340 nm

shipped in

wet ice

SMILES string

O.[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H29N7O14P2.2Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;;1H2/q;2*+1;/p-2/t10-,11-,13-,14-,15-,16-,20-,21-;;;/m1.../s1

InChI key

FWBNCIFELNNJCX-UHOVGGJYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.
NADH has been used in MDH (malate dehydrogenase) enzymatic activity assay.
NADH has been used in lactate dehydrogenase (LDH) enzymatic activity assay, glutamate-cysteine ligase activity assay and fructose 2,6-bisphosphate assay.

Biochem/physiol Actions

NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, β-oxidation and the citric acid cycle (Krebs cycle, TCA cycle). It participates in cell signaling events as well, for example as a substrate for the poly (ADP-ribose) polymerases (PARPs) during the DNA damage response. The NAD+/NADH dependent sirtuins play key roles in stress responses during events involving energy metabolism, with implications in cancer biology, diabetes and neurodegenerative disease.
As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.

Packaging

Packaged by solid weight.

Specifications

Absorbance coefficients: 14.3 x 103 l x mol-1 x cm-1 at 260 nm, 6.18 x 103 l x mol-1 x cm-1 at 334 nm (Hg), 6.3 x 103 l x mol-1 x cm-1 at 340 nm, 3.4 x 103 l x mol-1 x cm-1 at 365 nm (Hg)

Formula:
C21H27N7O14P2Na2

pH value in water: 7.5 ± 0.5 (100 mg/ml)

Physical form

Amorphous powder

Preparation Note

Activator: Mg2+ (5 to 10 mM)
Storage conditions (working solution): 10 mg/ml in Na-bicarbonate buffer, pH 9, contains at least 95% NADH after 4 weeks at 2 to 8 °C

Storage and Stability

Store at 2 to 8 °C. (Store dry!)

Analysis Note

Purity: 98% β-NADH-Na2 based on dry weight (enzymatically and absorbance at 340 nm), 99% (absorbance at 260 nm), A260 nm-A340 nm: <2.40
Organic solvents: <4% (GC)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José L Reyes et al.
Plant, cell & environment, 31(12), 1781-1790 (2008-09-03)
In plants, Late Embryogenesis Abundant (LEA) proteins typically accumulate in response to low water availability conditions imposed during development or by the environment. Analogous proteins in other organisms are induced when exposed to stress conditions. Most of this diverse set
Tardigrades use intrinsically disordered proteins to survive desiccation.
Boothby T C, et al.
Molecular Cell, 65(6), 975-984 (2017)
MAPK14/p38?-dependent modulation of glucose metabolism affects ROS levels and autophagy during starvation.
Desideri E, et al.
Autophagy, 10(9) (2014)
Glutathione participates in the modulation of starvation-induced autophagy in carcinoma cells.
Desideri E
Autophagy, 8(12), 1769-1781 (2012)
Michela Carraro et al.
Cell reports, 32(9), 108095-108095 (2020-09-03)
The mitochondrial permeability transition pore (PTP) is a Ca2+-activated channel that plays a key role in cell death. Thiol oxidation facilitates PTP opening, yet the targets and molecular mechanisms still await a definition. Here, we investigate the role of C141

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service