Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N2915

Sigma-Aldrich

PYR-41

≥98% (HPLC), powder

Synonym(s):

4[4-(5-Nitro-furan-2-ylmethylene)-3,5-dioxo-pyrazolidin-1-yl]-benzoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N3O7
CAS Number:
Molecular Weight:
371.30
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

red to brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CCOC(=O)c1ccc(cc1)N2NC(=O)\C(=C/c3ccc(o3)[N+]([O-])=O)C2=O

InChI

1S/C17H13N3O7/c1-2-26-17(23)10-3-5-11(6-4-10)19-16(22)13(15(21)18-19)9-12-7-8-14(27-12)20(24)25/h3-9H,2H2,1H3,(H,18,21)/b13-9+

InChI key

ARGIPZKQJGFSGQ-UKTHLTGXSA-N

Application

PYR-41 has been used as E1 inhibitor:
  • to determine the cellular degradation pathways on adeno-associated viruses (AAV) transduction
  • to assess its selectivity and potency by matrix assisted laser desorption/ionization-time of flight (MALDI-TOF) E2/E3 assay
  • to treat fully differentiated C2C12 myotubes in high glucose medium for determining its effect on protein ubiquitination and cell viability
  • in the pretreatment of human brain microvascular endothelial cells (hBMECs) for the inhibition studies before infection with bacteria

Biochem/physiol Actions

PYR-41 inhibits E1 ubiquitin ligases like mouse double minute 2 homolog (MDM2), Itchy (ITCH) and HOIL-1L interacting protein (HOIP). It has anticancer potential.
PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Heterogeneity in pneumolysin expression governs the fate of Streptococcus pneumoniae during blood-brain barrier trafficking
Surve MV, et al.
PLoS Pathogens, 14(7), e1007168-e1007168 (2018)
Sandhya Manohar et al.
Antioxidants & redox signaling, 31(15), 1133-1149 (2019-09-05)
Aims: Ubiquitin is a highly conserved protein modifier that heavily accumulates during the oxidative stress response. Here, we investigated
Inês B Santarino et al.
Scientific reports, 7(1), 5812-5812 (2017-07-21)
Erythrophagocytosis, the phagocytic removal of damaged red blood cells (RBC), and subsequent phagolysosome biogenesis are important processes in iron/heme metabolism and homeostasis. Phagolysosome biogenesis implies the interaction of nascent phagosomes with endocytic compartments and also autophagy effectors. Here, we report
The MALDI-TOF E2/E3 ligase assay as universal tool for drug discovery in the ubiquitin pathway
De Cesare V, et al.
Cell Chemical Biology, 25(9), 1117-1127 (2018)
Chemical modulation of endocytic sorting augments adeno-associated viral transduction
Berry GE and Asokan A
The Journal of Biological Chemistry, 291(2), 939-947 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service