Skip to Content
MilliporeSigma

MAK545

Beta-Lactamase Activity Assay Kit

sufficient for 200 colorimetric tests

Synonym(s):

β-Lactamase Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41116104
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


detection method

colorimetric

relevant disease(s)

immunological diseases

shipped in

wet ice

storage temp.

−20°C

General description

β-Lactamases are a large family of enzymes capable of hydrolyzing β-lactams. A β-Lactam ring is the common element in all beta-lactam antibiotics including penicillin derivatives, cephalosporins, monobactams, and carbapenems. Through hydrolysis, β-lactamase breaks the β-lactam ring open, deactivating the antibacterial properties. Bacteria from clinical and non-clinical settings are becoming increasingly resistant to β-lactam antibiotics by synthesizing β-lactamase. To overcome this resistance, β-lactam antibiotics are often given with β-lactamase inhibitors such as clavulanic acid. Therefore, detection of β-lactamase activity is of central importance to assess beta-lactam antibiotics as well as to prevent antibiotics resistance.

The Beta-Lactamase Activity Assay Kit offers a sensitive colorimetric assay for measuring β-lactamase activity in biological samples. The assay can be performed using an absorbance microplate reader by measuring the OD ratio at the wavelength of 490 nm to 380 nm.

Biochem/physiol Actions

The β-lactamase activity is detected using nitrocefin, which changes color from yellow to red upon hydrolysis by β-lactamase.



One unit of β-lactamase is the amount of enzyme required to hydrolyze 1.0 µmole of nitrocefin per minute at pH 7.0 at 25°C.


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library