Key Documents
MTCF4062F
MTC Bio BioFlask Erlenmeyer Flask
capacity 500 mL, flat PETG bottom, cap, vented, sterile, pack of 12 ea (individually wrapped)
Synonym(s):
culture flask
About This Item
UNSPSC Code:
41122104
NACRES:
NB.15
material
flat PETG bottom
polypropylene
sterility
sterile
feature
cap vented
packaging
pack of 12 ea (individually wrapped)
manufacturer/tradename
MTC Bio F4062-F
capacity
500 mL
General description
Flask, Erlenmeyer, 500ml, PETG, flat bottom w / vented screw cap, Sterile, indiv wrapped, 12/CS
Jumbo Fernbach Flasks made of styrene-butadiene with polypropylene screw caps which are vented with 0.2µm membranes to increase air transmission and enhance cell growth. The mouth size conforms to the international SBS standard, for quick, secure connection to bioreactor equipment.
Features and Benefits
  • Vented screw caps
  • Flat or baffled bottoms
  • Gamma sterile (SAL 10-6)
  • Pyrogen free and RNase/DNase free
  • Individually wrapped
Legal Information
BioFlask is a trademark of MTC Bio
MTC is a trademark of MTC Bio