Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.46
UNSPSC Code:
12352203
biological source
human (normal adult)
Quality Segment
General description
Tissue Section Slide – I, contains multiple paraffin embedded sections prepared from different human normal tissues. Each slide contains sections from 8 human adult normal tissues: Heart, Brain, Kidney, Liver, Lung, Pancreas, Spleen, Skeletal Muscle
Application
Suitable for immunohistochemistry and in situ hybridization. May be used for cellular localization of tissue specific mRNA and protein expression.
Tissue Section Slide – I is suitable for immunohistochemistry applications and has been used as a immunohistochemistry control slide. The slide may also be used for in situ hybridization and in situ PCR studies, which includes tissue specific protein and mRNA expression, to compare the location of novel genes among different tissues.
Preparation Note
Paraffin embeded tissue sections, approximately 5 μm thick.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Tissue slide
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.