Skip to Content
MilliporeSigma

MSQC17

SILuLite SigmaMAb Rituximab Monoclonal Antibody

recombinant, expressed in CHO cells

Synonym(s):

Mass spectrometry standard, Rituximab

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
41105331
Clone:
-
Species reactivity:
-
Application:
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in CHO cells

Quality Segment

assay

≥90% (SDS-PAGE)

form

solid

suitability

suitable for mass spectrometry

shipped in

wet ice

storage temp.

−20°C

General description

SigmaMAb Rituximab (MSQC17) was designed to be a standard for the analysis of Rituximab monoclonal antibodies using mass spectrometry. A heavy labeled version is available as SILuMab Rituximab (MSQC13).

SigmaMAb Rituximab is for R&D use only. Not for drug, household, or other uses.

Application

SILu Lite SigmaMAb Rituximab Monoclonal Antibody has been used to study its anticancer effects by antibody-dependent cell-mediated cytotoxicity (ADCC) mechanism.

Other Notes

SigmaMab Rituximab Heavy Chain:
QVQLQQPGAELVKPGASVKMSCKASGYTFTSYNMHWVKQTPGRGLEWIGAIYPGNGDTSYNQKFKGKATLTADKSSSTAYMQLSSLTSEDSAVYYCARSTYYGGDWYFNVWGAGTTVTVSAASTKGPSVFPLAPSSKSTSGGTAALGCLVKDYFPEPVTVSWNSGALTSGVHTFPAVLQSSGLYSLSSVVTVPSSSLGTQTYICNVNHKPSNTKVDKKAEPKSCDKTHTCPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVMHEALHNHYTQKSLSLSPG

SigmaMab Rituximab Light Chain:
QIVLSQSPAILSASPGEKVTMTCRASSSVSYIHWFQQKPGSSPKPWIYATSNLASGVPVRFSGSGSGTSYSLTISRVEAEDAATYYCQQWTSNPPTFGGGTKLEIKRTVAAPSVFIFPPSDEQLKSGTASVVCLLNNFYPREAKVQWKVDNALQSGNSQESVTEQDSKDSTYSLSSTLTLSKADYEKHKVYACEVTHQGLSSPVTKSFNRGEC

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library