MilliporeSigma
Functional and structural microanatomy of the fetal sciatic nerve.
Muscle & nerve (2016-12-23)
Maud Creze, Mazen Zaitouna, Nyangoh Timoh Krystel, Djibril Diallo, Cédric Lebacle, Marie-France Bellin, Denis Ducreux, Gérard Benoit, Thomas Bessede
ABSTRACT
The ultrastructure of a nerve has implications for surgical nerve repair. The aim of our study was to characterize the fascicular versus fibrillar anatomy and the autonomic versus somatic nature of the fetal sciatic nerve (SN). Immunohistochemistry for vesicular acetylcholine transporter, tyrosine hydroxylase, and peripheral myelin protein 22 was performed to identify cholinergic, adrenergic, and somatic axons, respectively, in the human fetal SN. Two-dimensional (2D) analysis and 3D reconstructions were performed. The fetal SN is composed of one-third stromal tissue and two-thirds neural tissue. Autonomic fibers are predominant over somatic fibers within the neural tissue. The distribution of somatic fibers is initially random, but then become topographically organized after intra- and interfascicular rearrangements have occurred within the nerve. The fetal model presents limitations but enables illustration of the nature of the nerve fibers and the 3D fascicular anatomy of the SN. Muscle Nerve 56: 787-796, 2017.
Sigma-Aldrich
Mayer′s Hematoxylin Solution
Pricing and availability is not currently available.