Skip to Content
MilliporeSigma
All Photos(1)

Documents

MGECKO2A

Sigma-Aldrich

Gecko2 Mouse Whole Genome CRISPR Pool, All-in-One Lenti Particles (Gecko2 vector)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

packaging

pkg of 8x25 μL (vials)

Quality Level

concentration

5x108  VP/ml (via p24 assay)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

General description

Developed by Feng Zhang′s lab at the Broad Institute, the mouse GeCKO v2 (all-in-one vector) libraries consist of over 100,000 unique gRNAs for gene knock-out in the mouse genome. Using an all-in-one lenti CRISPR vector which co-expresses Cas9 , gRNA with a puromycin selection marker, the version 2 vector produces nearly 10X higher titer than version 1. Sigma′s lentiviral mouse GeCKO pool all-in-one is provided in 8 x 25 ul aliquots at a minimum titer of 5X10^8 TU/ml (measured by a p24 assay).

Each species-specific library is delivered as two half-libraries (A and B). It is recommended to screen both A and B libraries together, which will include 6 sgRNAs per gene (3 sgRNAs in each library). Both libraries contain 1000 non-targeting control sgRNAs. The A library also targets miRNAs (4 sgRNAs per miRNA).

Application

Functional Genomics/Screening /Target Validation

Features and Benefits

  • Use CRISPR nucleases to knockout protein-coding genes to assess their function
  • Efficiently screen the whole human genome (16,000+ genes) at the bench-top without robotics or specialized equipment

Numerous built-in enrichment and depletion controls allow researchers to confidently gauge the success of their pooled screening experiments
  • Use the one vector system for the mouse GeCKO version 2 libraries in applications where the Cas9 cannot be introduced into mouse cells (like in primary cells).
  • Lentiviral CRISPRs can infect a broad variety of mammalian cells by co-expressing a mammalian codon-optimized Cas9 nuclease along with a single guide RNA (sgRNA) to facilitate gene knockout.
  • Use puromycin to select after transduction.

Preparation Note

Puro Kill Curve and Determining CFU (Colony Formation Unit) per mL. Prior to performing a library-scale screening, two preliminary experiments must be conducted. Visit Sigma.com/pooledscreening.

Other Notes

This product is for R&D use only, not for drug, household, or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices. Though the lentiviral transduction particles produced are replication incompetent, it is recommended that they be treated as Risk Group Level 2 (RGL-2) organisms in laboratory handling. Follow all published RGL-2 guidelines for laboratory handling and waste decontamination.

Legal Information

All-in-One is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This screening guide covers how to choose a cell line, a screening library, and experimental conditions as well as tips for designing and performing your experiment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service