Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRISPRILIB

Sigma-Aldrich

Human Whole Genome and Long Non Coding CRISPRi Library Kit

Synonym(s):

CRISPRi Library, Human Genome CRISPRi Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105904
NACRES:
NE.02

packaging

pkg of 80 vials (4x50μL aliquots for each of the 20 kit components)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

General description

CRISPRi is a potent tool for modulating gene expression. This technology uses dCas9 to act as a synthetic transcription factors by recruiting endogenous transcription repressor complexes to gene promoters and enhancers, resulting in down regulated gene transcription. CRISPRi achieves LOF phenotypes without the limitations of RNAi and CRISPR KO, although CRISPRi relies on delivery by lentivirus, this technology offers new possibilities for genome engineering. CRISPRi can be used individually to target areas of the genome that are inaccessible by other gene perturbation technologies (e.g., non-coding regions) or in conjunction to uncover gene-regulatory networks underlying discrete phenotypes. Finally, pooled CRISPRi screening can be paired with single-cell RNA-sequencing methods to enable high-dimensional characterization of CRISPR gene perturbation.

Human Whole Genome and Long Non Coding CRISPRi Library Kit contains 17 sub-pools of top 5 gRNAs per gene as well as a separate sub-pool of 5 supplemental gRNAs per gene for increased sensitivity and includes non-targeting controls built in. All pools are functionally titered based on # of guides to provide approximately 3 replicates depending on cell line at 500 gRNAs per cell. Vectors contain both BFP and puromycin as selection markers and an optimized scaffold for sgRNA expression and function. Each sub pool of the library includes non-targeting sgRNA controls. The UCOE KRAB-dCas9 effector is designed for consistent expression and inhibition allowing gene silencing loss-of-function screening with less risk of toxicity to your cells due to double stranded breaks which make the CRISPRi system the ideal choice for perturbation of non-coding genes, applications that call for a reversible, titratable solution, and essential genes.
  • Individual CRISPRi clones are easily ordered online or by contacting your local sales representative
  • Custom pools for follow up screening or 10x Genomics Compatible CRISPR pools are also available by contacting your local sales representative

For more information about screening with CRISPRi/a please Click Here.

Application

  • Functional Genomics/Target Validation
  • Unbiased wholed genome forward genetic screening
  • Set up and optimization of screen assay
  • Creation of cell lines stably expressing KRAB-dCas9
  • Validated positive and negative controls

Features and Benefits

  • Complete ready to use whole genome CRISPRi libarary targeting protein coding and long non-coding RNAs
  • Best in Class UCSF gRNA selection algorithm and optimized (F+E) gRNA scaffold
  • Ease of optimization: Validated positive control targeting RAB1A and non-targeting control for assay set up
  • UCOE KRAB-dCas9 for consistent effector expression across a wide variety of cell lines
  • Libraries provided at high functional titer, based on FACs or CFU analysis
  • Use puromycin and or BFP as selection markers
  • Gene targets from UCSF organized into functional categories, both coding and lncRNA Kinases, Phosphatases, Drug Targets Cancer and Apoptosis Stress and Proteostasis Mitochondria, Trafficking, Motility Gene Expression Membrane Proteins, Unassigned (Novel Targets)

Components

17 Subpools with a minimum concentration of 1x108 TU/mL(via FACS assay)
CRISPRIL1: CRISPRi Pool H1 Top5 Kinase Phosphatase Drug Targets
CRISPRIL2: CRISPRi Pool H1 Supp5 Kinase Phosphatase Drug Targets
CRISPRIL3: CRISPRi Pool H2 Top5 Cancer Apoptosis
CRISPRIL4: CRISPRi Pool H2 Supp5 Cancer Apoptosis
CRISPRIL5: CRISPRi Pool H3 Top5 Stress Proteostasis
CRISPRIL6: CRISPRi Pool H3 Supp5 Stress Proteostasis
CRISPRIL7: CRISPRi Pool H4 Top5 Mitochondria Trafficking Motility
CRISPRIL8: CRISPRi Pool H4 Supp5 Mitochondria Trafficking Motility
CRISPRIL9: CRISPRi Pool H5 Top5 Gene Expression
CRISPRIL10: CRISPRi Pool H5 Supp5 Gene Expression
CRISPRIL11: CRISPRi Pool H6 Top5 Membrane Proteins
CRISPRIL12: CRISPRi Pool H6 Supp5 Membrane Proteins
CRISPRIL13: CRISPRi Pool H7 Top5 Unassigned
CRISPRIL14: CRISPRi Pool H7 Supp5 Unassigned
CRISPRIL15: CRISPRi Pool Long Noncoding iPSC and Common Cancer Cell Lines 1
CRISPRIL16: CRISPRi Pool Long Noncoding iPSC 13
CRISPRIL17: CRISPRi Pool Long Noncoding Common Cancer Cell Lines 2

2 Controls and 1 Effector Construct with a minimum concentration of 5x105 TU/mL(via FACS or CFU assay)
CRISPRIE: KRAB-dCAS9 CRISPRi Construct Lentiviral Transduction Particles
CRISPRI01: CRISPRi Rab1a Control Lentiviral Transduction Particles
CRISPRI06: CRISPRi Nontargeting Control Puromycin Transduction Particles

Principle

The power of CRISPR for genome engineering, coupled with the ability to perform large-scale, whole genome, loss-of-function (LOF) screening, has allowed for new breakthroughs identifying gene pathways in drug resistance and disease. CRISPR is most commonly used to create double-stranded breaks that often result in loss of gene function (CRISPR-KO). However, the full extent of CRISPR′s utility extends beyond just targeted cutting of DNA. Nuclease-independent applications of CRISPR provide equal targeting specificity but instead of cutting, CRISPRi allows for targeted interference of gene function by delivering transcriptional repressor domains to a specific target sequence using modified dCas9 + gRNA complexes. Gene knockdown is complementary to CRISPR-KO and CRISPRa (activation) and has distinct advantages over existing loss-of-function strategies like RNAi. We partnered with University of California San Francisco to provide the best-in-class CRISPRi screening tools available.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CRISPR lentiviral screening libraries, partnered with 10x Genomics, offer powerful research tools for pooled screening.

CRISPR lentiviral screening libraries, partnered with 10x Genomics, offer powerful research tools for pooled screening.

CRISPR lentiviral screening libraries, partnered with 10x Genomics, offer powerful research tools for pooled screening.

CRISPR lentiviral screening libraries, partnered with 10x Genomics, offer powerful research tools for pooled screening.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service