Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MBD0008

Sigma-Aldrich

Inactivated Proteus vulgaris

Suitable for DNA extraction, PCR, sequencing, next generation sequencing, >10^8 bacteria/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.51

Quality Level

form

liquid

concentration

>10^8 bacteria/mL

technique(s)

DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses.1 Inactivated Proteus vulgaris can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs.
Proteus vulgaris is a rod-shaped gram negative, facultative anaerobe bacterium. It inhabits the intestinal tract of humans and animals and can be found in soil, water and feces.2 P. vulgaris is a member of Enterobacteriacea family that are opportunistic pathogens in humans, responsible for urinary tract and burn infections. 3 P. vulgaris was also observed fecal samples of healthy individuals. 4

Read here how to use our standards to ensure data integrity for your microbiome research.


Application

Inactivated bacteria are provided at >10^8 bacteria/ml concentration in TE buffer pH 8.0. It is recommended to avoid freeze thaw cycles of this product.
Suitable for Quantitative standard for PCR, Sequencing and NGS

Features and Benefits

  • Individual microbial standard for microbiomics and meta-genomics workflow
  • Suitable standard for PCR, sequencing and NGS
  • Improve Bioinformatics analyses
  • Increases reproducibility
  • Compare results lab to lab

Physical form

Liquid -The inactivated bacteria is provided at >10^8 bacteria/ml in TE buffer pH 8.0

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service