Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK508

Sigma-Aldrich

Choline Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

food and beverages
pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

anxiety; neurological disorders

shipped in

wet ice

storage temp.

−20°C

General description

Choline and its metabolites play important roles in membrane structure integrity, cellular signaling and cholinergic neurotransmission. Aberrant regulation in choline metabolism has been associated with mental illness such as anxiety.

Features and Benefits

  • Linear detection range: 1 to 100 µM for the colorimetric assay and 0.2 to 10 µM for the fluorometric assay.

Suitability

The kit is suitable for the determination of choline in biological samples such as serum, plasma, urine, saliva, milk, tissue, and cell culture, as well as for studying the effects of drugs on choline metabolism.

Principle

The Choline Assay Kit method provides a simple, direct and high-throughput assay for measuring choline in biological samples. In this assay, free choline is oxidized by choline oxidase to betaine and H2O2 which reacts with a specific dye to form a pink colored product. The color intensity at 570 nm or fluorescence intensity λEx = 530 nm/λEm = 585 nm is directly proportional to the choline concentration in the sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service