Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK465

Sigma-Aldrich

Glycogen Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Glycogen Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

diabetes; endocrinological disorders

shipped in

wet ice

storage temp.

−20°C

General description

Glycogen is a branched polysaccharide of glucose units linked by α-1,4 glycosidic bonds and α-1,6 glycosidic bonds. It is stored primarily in the liver and muscle and forms an energy reserve that can be quickly mobilized to meet a sudden need for glucose. The most common glycogen metabolism disorder is found in diabetes, in which liver glycogen can be abnormally accumulated or depleted due to abnormal amounts of insulin. Genetic glycogen storage diseases have been associated with various inborn errors of metabolism caused by deficiencies of enzymes necessary for glycogen synthesis or breakdown.

Features and Benefits

  • Linear detection range: 2 to 200 µg/mL glycogen for colorimetric assays and 0.2 to 20 µg/mL for fluorometric assays.
  • Convenient room temperature assay. The procedure involves adding a single working reagent, and and takes only 30 minutes

Suitability

The kit is suitable for the quantitative determination of glycogen in biological samples such as tissue and cells, as well as the evaluation of drug effects on glycogen metabolism.

Principle

The Glycogen Assay Kit uses a single working reagent that combines the enzymatic break down of glycogen and the detection of glucose in one step. The color intensity of the reaction product at 570 nm or fluorescence intensity at λEx  = 530 nm/λEm = 585 nm is directly proportional to the glycogen concentration in the sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service