Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

M9027

Sigma-Aldrich

MOPS hemisodium salt

≥99% (titration)

Synonym(s):

3-(N-Morpholino)propanesulfonic acid hemisodium salt, 4-Morpholinepropanesulfonic acid hemisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14.5NO4S · 0.5Na
CAS Number:
Molecular Weight:
220.25
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (titration)

form

powder or crystals

useful pH range

6.5-7.9

pKa (25 °C)

7.2

solubility

water: 500 mg/mL, clear, colorless to almost colorless

SMILES string

[Na+].OS(=O)(=O)CCCN1CCOCC1.[O-]S(=O)(=O)CCCN2CCOCC2

InChI

1S/2C7H15NO4S.Na/c2*9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h2*1-7H2,(H,9,10,11);/q;;+1/p-1

InChI key

OVCWZKQPQOVIDT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service