Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7024

Sigma-Aldrich

Murashige and Skoog Shoot Multiplication Medium A

powder, suitable for plant cell culture

Synonym(s):

MSMA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.72

form

powder

Quality Level

technique(s)

cell culture | plant: suitable

application(s)

agriculture

shipped in

ambient

storage temp.

2-8°C

Application

Murashige and Skoog Shoot Multiplication Medium A is a widely used plant tissue culture growth medium. M&S Basal Medium contains macronutrients that include high levels of nitrate and organic additives such as agar, sugars, vitamins and growth regulators. Important growth regulators frequently added to M&S include IAA (auxin/morphogen) and the Kinetin (cytokinin/cell division promoter).

Formula variant

With the macro- and micronutrients, vitamins, sucrose, IAA, sodium phosphate, adenine and 2iP as described by Huang and Murashige (1976) TCA Manual.

Media Formulation

Preparation Note

Murashige and Skoog medium can be reconstituted from powder or by combining products that are major components of complete M&S medium, such as macronutrient mixtures and vitamin mixtures. Murashige and Skoog Shoot Multiplication Medium A (M7024 is a variant media) with macro- and micronutrients, vitamins, sucrose, IAA, sodium phosphate, adenine and 2iP.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plant tissue culture media: major constituents, their preparation and some applications.
Huang L, Murashige T.
Tissue Antigens, 3, 539-548 (1976)
R H Smith et al.
In vitro, 13(5), 329-334 (1977-05-01)
Defined in vitro conditions for callus initiation by Gossypium arboreum L. were determined, and different tissues were evaluated as explant sources, Environmental conditions tested included light versus dark, and low light versus high light. Different nutrient media as well as
Akasaka et al.
Plant science : an international journal of experimental plant biology, 156(2), 169-175 (2000-08-11)
Bud primordia were induced from leaf segments, which were harvested from young seedlings of Spanish type peanut (Arachis hypogaea L. cv. Chico), on 0.8% agar-solidified medium containing Murashige and Skoog (MS) basal salts supplemented with B5 vitamins, 1 mg/l NAA
Regulation of lycopene formation in cell suspension culture of VFNT tomato (Lycopersicon esculentum) by CPTA, growth regulators, sucrose, and temperature
Robertson GE, Mahoney NE
Journal of Experimental Botany, 46, 667-673 (1995)
Nor Azlina Hasbullah et al.
Pakistan journal of biological sciences : PJBS, 11(11), 1449-1454 (2008-09-27)
Regeneration potentials in Gerbera jamesonii Bolus ex. Hook f. from tissues culture system was studied using leaf, petiole and root explants. In vitro regeneration, callus induction and root formation were optimized by manipulation of growth regulators during organogenesis. Various kinds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service