Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2727

Sigma-Aldrich

Mexiletine hydrochloride

≥98% (GC), powder, sodium channel blocker

Synonym(s):

1-(2,6-Dimethylphenoxy)-2-propanamine hydrochloride, 1-(2,6-Xylyloxy)-2-aminopropane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO · HCl
CAS Number:
Molecular Weight:
215.72
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Mexiletine hydrochloride, powder

Assay

≥98% (GC)

form

powder

color

white to off-white

solubility

methanol: 50 mg/mL

originator

Boehringer Ingelheim

storage temp.

2-8°C

SMILES string

Cl[H].CC(N)COc1c(C)cccc1C

InChI

1S/C11H17NO.ClH/c1-8-5-4-6-9(2)11(8)13-7-10(3)12;/h4-6,10H,7,12H2,1-3H3;1H

InChI key

NFEIBWMZVIVJLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Mexiletine is a class I B antiarrhythmic and an analog of lidocaine. It has shelf life of 10-12 hours and is metabolized in liver and eliminated post reduction, oxidation deamination or conjugation.

Application

Mexiletine hydrochloride has been used as a sodium channel blocker:
  • expressed in chinese hamster ovary cells
  • in human embryonic kidney (HEK) cells for whole cell patch-clamp studies
  • electrophysiology studies in HEK cells expressing Nav1.7 protein

Biochem/physiol Actions

Mexiletine is a potent sodium channel blocker. It is a cardiac antiarrhythmic and is used as an adjuvant in headache and neuropathic pain Mexiletine is used for treating myotonia in sodium channelopathies and reduces the cardiac action potential depolarization but shows no impact on atrial refractoriness. Its inhibitory effect on sodium channels is effective in treating potassium aggravated myotonia.

Features and Benefits

This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnold E Pfahnl et al.
Heart rhythm, 4(1), 46-53 (2007-01-03)
Brugada and long QT type 3 syndromes are linked to sodium channel mutations and clinically cause arrhythmias that lead to sudden death. We have identified a novel threonine-to-isoleucine missense mutation at position 353 (T353I) adjacent to the pore-lining region of
E L Logigian et al.
Neurology, 74(18), 1441-1448 (2010-05-05)
To determine if mexiletine is safe and effective in reducing myotonia in myotonic dystrophy type 1 (DM1). Myotonia is an early, prominent symptom in DM1 and contributes to decreased dexterity, gait instability, difficulty with speech/swallowing, and muscle pain. A few
J F Desaphy et al.
Neuromuscular disorders : NMD, 9(3), 182-189 (1999-06-26)
The sea anemone toxin ATX II impairs skeletal muscle sodium channel inactivation, mimicking the persistent inward current observed in patients suffering from sodium channel myotonia. Mexiletine has beneficial effects on myotonia. To verify the efficiency of the drug on persistent
Min-Tzu Wu et al.
PloS one, 8(1), e55212-e55212 (2013-02-06)
Primary erythromelalgia (PE) is an autosomal dominant neurological disorder characterized by severe burning pain and erythema in the extremities upon heat stimuli or exercise. Mutations in human SCN9A gene, encoding the α-subunit of the voltage-gated sodium channel, Na(v)1.7, were found
Erythromelagia: a rare and hard-to-treat condition: a 9-year-old boy responsive to intravenous lidocaine and oral mexilitene.
Francisca Elgueta et al.
Pain medicine (Malden, Mass.), 14(2), 311-312 (2013-02-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service