Skip to Content
MilliporeSigma
Mexiletine hydrochloride European Pharmacopoeia (EP) Reference Standard

Mexiletine hydrochloride

Mexiletine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-(2,6-Dimethylphenoxy)-2-propanamine hydrochloride, 1-(2,6-Xylyloxy)-2-aminopropane hydrochloride, Mexiletine hydrochloride
Empirical Formula (Hill Notation):
C11H17NO · HCl
CAS Number:
Molecular Weight:
215.72

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade