Documents
M2683
Sigma-Aldrich
Media bottles, wide mouth, Lab 45
capacity 500 mL
Synonym(s):
bottle wide mouth, wide mouth bottle, wide mouth bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
borosilicate glass bottle
polypropylene cap
Joint
Joints threaded (45)
description
Autoclavable
sterility
non-sterile
feature
graduations
with cap
packaging
pack of 12 ea
manufacturer/tradename
WHEATON®
capacity
500 mL
General description
Graduated safety coated bottles are ideal for storing fluids, dry chemicals, and other materials in the lab and help protect against broken glass. The strong, slip resistant plastisol coating reduces potential breakage and exposure to lab personnel handling hazardous material. Manufactured from Wheaton glass (meets USP Type I standards). Fluids are protected because the glass has a low alkali content. Wide opening, special lip and sloping shoulders facilitate pouring and cleaning. Includes a white polypropylene screw cap with a wedge-shaped inner sealing ring and polypropylene pour ring.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.