Skip to Content
MilliporeSigma

M2808

Media bottles, wide mouth, Lab 45

capacity 1000 mL

Synonym(s):

bottle wide mouth, wide mouth bottle, wide mouth bottles

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.17
UNSPSC Code:
41121800
Capacity:
1000 mL
Material:
borosilicate glass bottle, polypropylene cap, round bottle
Sterility:
non-sterile
Feature:
graduations, with cap
Packaging:
pack of 12 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

borosilicate glass bottle, polypropylene cap, round bottle

joint

Joints threaded neck (45)

description

Autoclavable

sterility

non-sterile

feature

graduations, with cap

packaging

pack of 12 ea

manufacturer/tradename

WHEATON® 219940

capacity

1000 mL

General description

Graduated safety coated bottles are ideal for storing fluids, dry chemicals, and other materials in the lab and help protect against broken glass. The strong, slip resistant plastisol coating reduces potential breakage and exposure to lab personnel handling hazardous material. Manufactured from Wheaton glass (meets USP Type I standards). Fluids are protected because the glass has a low alkali content. Wide opening, special lip and sloping shoulders facilitate pouring and cleaning. Includes a white polypropylene screw cap with a wedge-shaped inner sealing ring and polypropylene pour ring.

Features and Benefits

  • Plastisol coated exterior of bottle retains contents if bottle is accidentally broken
  • Made from low extractable clear borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • 45 mm white polypropylene screw cap attached
  • Pour ring eliminates drips
  • Autoclavable
  • Color coded caps available separately

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.


Still not finding the right product?

Explore all of our products under Media bottles, wide mouth, Lab 45




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library