Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
material
black polystyrene , flat bottom black polystyrene wells (with micro-clear bottom)
sterility
sterile
feature
lid, skirt (F-bottom)
packaging
case of 32
manufacturer/tradename
Greiner 655090
maximum volume
392 μL
size
96 wells
working volume
340 μL
binding type
Tissue Culture (TC)-treated surface
General description
Application
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material black polystyrene , flat bottom black polystyrene wells (with micro-clear bottom) | material black polystyrene plate, flat bottom black polystyrene wells (chimney style) | material flat bottom clear wells (chimney style), white polystyrene plate | material black polystyrene , flat bottom clear polystyrene wells (with micro-clear bottom) |
| manufacturer/tradename Greiner 655090 | manufacturer/tradename Greiner 655086 | manufacturer/tradename Greiner 655098 | manufacturer/tradename Greiner 655087 |
| feature lid, skirt (F-bottom) | feature lid (with condensation rings), skirt (F-bottom) | feature lid (with condensation rings), skirt (F-bottom) | feature lid: no, skirt (F-bottom) |
| binding type Tissue Culture (TC)-treated surface | binding type Tissue Culture (TC)-treated surface | binding type Tissue Culture (TC)-treated surface | binding type Tissue Culture (TC)-treated surface |
| size 96 wells | size 96 wells | size 96 wells | size 96 wells |
Still not finding the right product?
Explore all of our products under Greiner CELLSTAR® 96 well plates
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
