Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M010800

Sigma-Aldrich

TFA-Hexylaminolinker Phosphoramidite

Synonym(s):

TFA-Hexylaminolinker Amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31F3N3O3P
CAS Number:
Molecular Weight:
413.42
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)

bulk density

1.095 g/mL

suitability

conforms to structure for H-NMR (Identity)

shipped in

dry ice

storage temp.

−70°C

SMILES string

CC(C)N(C(C)C)P(OCCCCCCNC(=O)C(F)(F)F)OCCC#N

InChI

1S/C17H31F3N3O3P/c1-14(2)23(15(3)4)27(26-13-9-10-21)25-12-8-6-5-7-11-22-16(24)17(18,19)20/h14-15H,5-9,11-13H2,1-4H3,(H,22,24)

InChI key

SMMKTKILBQHSFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service