Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE1336

Sigma-Aldrich

Anti-Wee1

from rabbit, purified by affinity chromatography

Synonym(s):

Wee1-like protein kinase, EC: 2.7.10.2, WEE1hu, Wee1A kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... WEE1(7465)

General description

Wee1-like protein kinase (UniProt: P30291; also known as EC: 2.7.10.2, WEE1hu, Wee1A kinase) is encoded by the WEE1 gene (Gene ID: 7465) in human. Wee1 is a magnesium-dependent nuclear kinase that belongs to the Serine/Threonine family of protein kinases. It serves as a negative regulator of entry into mitosis (G2 to M transition) by protecting the nucleus from cytoplasmically activated cyclin B1-complexed Cdk1 prior to the onset of mitosis. Two isoforms of Wee1 have been described that are produced by alternative splicing. In isoform 1 the nucleotide binding site is localized to amino acids 305-313 and its protein kinase domain lies within amino acids 299-569. Wee1 specifically phosphorylates and inactivates cyclin B1-complexed Cdk1, reaching a maximum during G2 phase and a minimum as cells enter into M phase. Phosphorylation of cyclin B1-Cdk1 is shown to occur exclusively on Tyrosine 15. Wee1 levels are believed to increase during S and G2 phases due to increase in transcription and a decline in M phase has been attributed to reduced synthesis and higher degradation. Phosphorylation at serine 642 by BRSK1 and BRSK2 in post-mitotic neurons is reported to down-regulate Wee1 activity in polarized neurons. Also, phosphorylation at serine 53 by PLK1 and at serine 123 by Cdk1 generates a signal for its ubiquitination and proteasomal degradation at the onset of G2/M phase.

Specificity

This rabbit polyclonal antibody detects Wee1-like kinase in human cells. It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Wee1-like protein kinase. The immunogen sequence is conserved between isoforms 1 and 2.

Application

Anti-Wee1, Cat. No. ABE1336-100UG, is a highly specific rabbit polyclonal antibody that targets Wee1-like protein kinase and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 2 µg/mL from a representative lot detected Wee1 in 10 µg of K562 cell lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Wee1 in human placenta and human testis cancer tissues.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Wee1 in 10 µg of HepG2 cell lysate.

Target description

~72 kDa observed; 71.60 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service