Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K10025A

Sigma-Aldrich

5′-Nucleotides & Nucleosides

Synonym(s):

Nucleoside Triphosphates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.51

form

powder

shipped in

dry ice

storage temp.

−20°C

General description

Nucleosides comprise the natural nucleobases, namely, cytosine (C), thymine (T), uracil (U), adenine (A), and guanine (G) with the sugar ring. DNA and RNA have 2′-deoxyribose and ribose respectively. Nucleotides have nucleoside and a minimum of one phosphate group. They are constituents of nucleic acids and are regarded as building blocks for the synthesis and in nanomaterials assembly as well.

Application

5′-Nucleotides & Nucleosides has been used as a standard mixture in gas chromatography using atmospheric pressure chemical ionization coupled to mass spectrometry (GC/APCI-MS) analysis. It has also been used as a nucleotide standard to generate a calibration curve in high-performance liquid chromatography (HPLC).

Components

16 Nucleotides & Nucleosides in quantities as indicated
A9251 Adenosine, 100mg
A2754 ADP, Sodium, 100mg
A1752 AMP, Sodium, 100mg
A2383 ATP, Disodium, 100mg
C4654 Cytidine, 100mg
C9755 CDP, Sodium, 25mg
C1006 CMP, Sodium, 25mg
C9274 CTP, Sodium, 25mg
G6752 Guanosine, 100mg
G7127 GDP, Sodium, 25mg
G8377 GMP, Sodium, 25mg
G8877 GTP, Sodium, 25mg
U3750 Uridine, 100mg
U4125 UDP, Sodium, 25mg
U6375 UMP, Disodium, 25mg
U6625 UTP, Sodium, 25mg

Other Notes

5′-Nucleotides & Nucleosides is a set of adenyl, guanyl, cytosyl and uridyl nucleotides and nucleosides that may be used in studies requiring full sets of these purines and pyrimidines.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Pu et al.
Chemical Society reviews, 47(3), 1132-1132 (2018-01-18)
Correction for 'Nucleobases, nucleosides, and nucleotides: versatile biomolecules for generating functional nanomaterials' by Fang Pu et al., Chem. Soc. Rev., 2018, DOI: .
S L Blanco López et al.
Journal of chromatography. A, 891(1), 99-107 (2000-09-22)
Reversed-phase HPLC was applied to obtain a sensitive and efficient means for quantitating nucleotides in the mussel Mytilus galloprovincialis. We obtained a good separation of adenylic, guanylic, uridylic and cytidylic nucleotides. Adenine nucleotides play a critical role in the regulation
Carsten Jaeger et al.
Analytical chemistry, 88(19), 9386-9390 (2016-09-02)
Gas chromatography using atmospheric pressure chemical ionization coupled to mass spectrometry (GC/APCI-MS) is an emerging metabolomics platform, providing much-enhanced capabilities for structural mass spectrometry as compared to traditional electron ionization (EI)-based techniques. To exploit the potential of GC/APCI-MS for more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service