Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47310-U

Supelco

Nucleosides Test Mix

in 1% sodium formate (varied conc.), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in 1% sodium formate (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

compatibility

use to QA LC-18-S columns

format

multi-component solution

separation technique

reversed phase

storage temp.

2-8°C

General description

Nucleosides Test Mix enables the troubleshooting of chromatographic problems, optimization of system efficiency, and evaluation of columns. These test mixes are shipped in amber ampules to prevent photodegradation. The instructions for proper use and interpretation of results are included. The Nucleosides Test Mix can be suitably chosen according to column-specific or application-specific requirements. The identification can be carried out using UV detection. For additional information, HPLC Troubleshooting Guide (Bulletin 826) may be requested.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Analyte

Description

    Cytidine 50 μg/mL
    Guanosine 25 μg/mL
    Inosine 25 μg/mL
    1-Methyladenosine 25 μg/mL
    5-Methylcytidine 100 μg/mL
    2′-O-Methylcytidine 20 μg/mL
    3-Methylcytidine methosulfate 100 μg/mL
    7-Methylguanosine 25 μg/mL
    5-Methyluridine 50 μg/mL
    β-Pseudouridine 25 μg/mL
    2-Thiocytidine dihydrate 10 μg/mL
    Uridine 25 μg/mL
    See All (12)

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

Chromatograms

application for HPLCapplication for HPLCapplication for LC-MSapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service