Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I5885

Sigma-Aldrich

Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse

clone GG-5, ascites fluid

Synonym(s):

Monoclonal Anti-Human IgG (γ-chain specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

GG-5, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:20,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Binds human IgG; does not bind other human Igs.The antibody is specific for a determinant on the heavy chain of human IgG. This antibody will form antigen-antibody complexes in the liquid phase in the presence of 3% PEG 6000.
Monoclonal Anti-Human IgG (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. Immunoglobulins have two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein. IgG is usually found as a monomer. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4. About 70 percent of the total immunoglobulin consists of IgG.

Application

Elisa assays were performed using alkaline phosphatase conjugated monoclonal mouse anti-human IgG to detect antibodies against various S. Aureus antigens in the serum from healthy individuals. The antibody was diluted 1:30000 in PBSt and incubated with samples for 2 hours at 37 degrees. Reactin was developed using p-nitrophenyl phosphate substrate (Sigma).
Liquid precipitate (Lppt).
Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse has been used in enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Immunoglobulin G (IgG) mainly helps in immune defense. IgG participates in hypersensitivity type II and type III.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alain Townsend et al.
Nature communications, 12(1), 1951-1951 (2021-03-31)
Serological detection of antibodies to SARS-CoV-2 is essential for establishing rates of seroconversion in populations, and for seeking evidence for a level of antibody that may be protective against COVID-19 disease. Several high-performance commercial tests have been described, but these
Utility of the P 19 suppressor of gene-silencing protein for production of therapeutic antibodies in N icotiana expression hosts
Garabagi F, et al.
Plant Biotechnology Journal, 10(9), 1118-1128 (2012)
Charles Pilette et al.
Immunology and cell biology, 88(4), 486-493 (2010-01-20)
As previously reported by others for immunoglobulin (Ig)G, we observed that IgA can induce interleukin (IL)-10 expression in human monocytes. In this study, we explored the molecular mechanisms of IL-10 induction by IgA in monocytes and monocyte-derived dendritic cells (MD-DCs).
Maha Zohra Ladjemi et al.
The European respiratory journal, 45(4), 980-993 (2014-12-30)
Despite their relevance to mucosal defense, production of IgA and the function of lung B-cells remain unknown in chronic obstructive pulmonary disease (COPD). We assessed IgA synthesis in the lungs of COPD (n=28) and control (n=21) patients, and regulation of
N Ruijne et al.
Clinical and vaccine immunology : CVI, 13(7), 797-801 (2006-07-11)
Vaccine trials with infants enrolled between 6 and 10 weeks of age (young infants) and 6 and 8 months of age (older infants) provided an opportunity to evaluate immunoglobulin G (IgG) isotype distribution and avidity maturation as indicators of antibody

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service